Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HKDC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566669
Description
Immunogen sequence: DLGGSKFRVLK VQVAEEGKRH VQMESQFYPT PNEIIRGNGT ELFEYVADCL ADFMKTKDLK HKK Highest antigen sequence identity to the following orthologs - mouse 84%, rat 89%.
The epidermal growth factor receptor (HKDC1; ErbB-1; HER1 in humans) is the cell-surface receptor for members of the epidermal growth factor family (EGF-family) of extracellular protein ligands. It is a member of the ErbB family of receptors, a subfamily of four closely related receptor tyrosine kinases: HKDC1 (ErbB-1), HER2/c-neu (ErbB-2), Her 3 (ErbB-3) and Her 4 (ErbB-4). HKDC1 exists on the cell surface and is activated by binding of its specific ligands, including epidermal growth factor and transforming growth factor alpha (TGFalpha). HKDC1 and its ligands are cell signaling molecules involved in diverse cellular functions, including cell proliferation, differentiation, motility, and survival, and in tissue development. Mutations that lead to HKDC1 overexpression (known as upregulation) or overactivity have been associated with a number of cancers, including lung cancer and glioblastoma multiforme. In this latter case a more or less specific mutation of HKDC1, called HKDC1vIII is often observed.
Specifications
HKDC1 | |
Polyclonal | |
Unconjugated | |
Hkdc1 | |
BC016235; hexokinase 1-like; hexokinase domain containing 1; hexokinase domain-containing protein 1; Hexokinase HKDC1; Hkdc1; LOC100364027; putative hexokinase HKDC1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80201 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q2TB90 | |
Hkdc1 | |
Recombinant Human HKDC1. Recombinant protein control fragment (Product #RP-107317). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction