Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA A Antibody (SR1692), Novus Biologicals™

Rabbit Monoclonal Antibody
$282.00 - $393.00
Specifications
Antigen | HLA A |
---|---|
Clone | SR1692 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Description
HLA A Monoclonal antibody specifically detects HLA A in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
HLA A | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
3105 | |
IgG | |
Affinity purified |
SR1692 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human | |
A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80 | |
A synthesized peptide derived from human HLA A (Uniprot #: P30443, P01889, P01891, P01892, P13746, P16188, P30450) | |
Primary | |
Store at -20 degrees C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title