Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA A2 Antibody (BB7.2), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP144896PEATT594
Description
HLA A2 Monoclonal specifically detects HLA A2 in Human samples. It is validated for Flow Cytometry.Specifications
HLA A2 | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
HLA-A | |
HLA-A2 solubilised by papain | |
0.1 mL | |
Primary | |
The antibody BB7.2 recognizes an epitope at the C-terminus of alpha-2 helix and a turn on one of the underlying beta strands within the human HLA-A2 histocompatibility antigen. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
BB7.2 | |
Flow Cytometry | |
A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80 | |
Mouse | |
Protein A purified | |
Adaptive Immunity, Cell Biology, Immunology | |
3105.0 | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction