Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HLA-A2 Monoclonal Antibody (BB7.2), PE, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 12987642
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-987-642 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-987-642 Supplier Invitrogen™ Supplier No. 12987642
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Description: This BB7.2 monoclonal antibody recognizes the human major histocompatibility complex (MHC) class I molecule HLA-A2, which is a member of the HLA-A family. MHC class I exists as a single alpha chain composed of three domains. This molecule associates with beta 2-microglobulin and is expressed on all nucleated cells. The BB7.2 monoclonal antibody has been reported to exhibit blocking activity, as well as recognize HLA-A28. Applications Reported: This BB7.2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This BB7.2 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser

HLA-A belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. Class I molecules play a central role in the immune system by presenting peptides derived from the endoplasmic reticulum lumen. They are expressed in nearly all cells. The heavy chain is approximately 45 kDa and its gene contains 8 exons. Exon 1 encodes the leader peptide, exons 2 and 3 encode the alpha1 and alpha2 domains, which both bind the peptide, exon 4 encodes the alpha3 domain, exon 5 encodes the transmembrane region, and exons 6 and 7 encode the cytoplasmic tail. Polymorphisms within exon 2 and exon 3 are responsible for the peptide binding specificity of each class one molecule. Typing for these polymorphisms is routinely done for bone marrow and kidney transplantation. Hundreds of HLA-A alleles have been described. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-A2
Applications Flow Cytometry
Classification Monoclonal
Clone BB7.2
Concentration 5 μL/Test
Conjugate PE
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene HLA-A
Gene Accession No. P04439
Gene Alias A29 protein; antigen presenting molecule; Aw-33; Aw-74; DAQB-90C11.16; FLJ26655; HLA A; HLA class I antigen; HLA class I histocompatibility antigen, A alpha chain; HLA class I histocompatibility antigen, A-1 alpha chain; HLA class I histocompatibility antigen, A-30 alpha chain; HLA class I histocompatibility antigen, A-33 alpha chain; HLA class I histocompatibility antigen, A-74 alpha chain; HLA locus; HLAA; HLA-A; HLA-A locus alpha 2 domain; HLA-A null; HLA-A11; HLA-A33; HLA-Aw33.1; HLA-DQB1; HLA-DRB1; Human leukocyte antigen A; leucocyte antigen A; leukocyte antigen class I-A; major histocompatibility complex, class I, A; MCH class I antigen; MHC 1; MHC class 1 antigen; MHC class I anti; MHC Class I antigen; MHC class I antigen A*30; MHC class I antigen A*33; MHC class I antigen A*74; MHC class I antigen heavy chain; MHC class I antigen HLA-A heavy chain; MHC class I antigen HLA-A33; MHC class I antigen null protein; MHC class I antigene; MHC class II antigen; MHC classI antigen; MHC1; truncated MHC class I antigen
Gene Symbols HLA-A
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3105
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.