Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA-A3 Monoclonal Antibody (GAP.A3), NovaFluor™ Blue 660-120S, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$201.00 - $430.00
Specifications
Antigen | HLA-A3 |
---|---|
Clone | GAP.A3 |
Concentration | 0.6 μg/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H071T03B08A
![]() |
Invitrogen™
H071T03B08A |
100 Tests |
Each for $430.00
|
|
|||||
H071T02B08A
![]() |
Invitrogen™
H071T02B08A |
25 Tests |
Each for $201.00
|
|
|||||
Description
HLA-A3 Monoclonal antibody specifically detects HLA-A3 in Human samples. It is validated for Flow CytometrySpecifications
HLA-A3 | |
0.6 μg/Test | |
Monoclonal | |
Mouse | |
Human | |
P04439 | |
3105 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
GAP.A3 | |
Flow Cytometry | |
NovaFluor Blue 660-120S | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
A29 protein; antigen presenting molecule; Aw-33; Aw-74; HLA A; HLA class I antigen; HLA class I histocompatibility antigen, A alpha chain; HLA class I histocompatibility antigen, A-1 alpha chain; HLA class I histocompatibility antigen, A-30 alpha chain; HLA class I histocompatibility antigen, A-33 alpha chain; HLA class I histocompatibility antigen, A-74 alpha chain; HLA locus; HLAA; HLA-A; HLA-A locus alpha 2 domain; HLA-A null; HLA-A11; HLA-A33; HLA-Aw33.1; HLA-DQB1; HLA-DRB1; Human leukocyte antigen A; leukocyte antigen class I-A; major histocompatibility complex, class I, A; MCH class I antigen; MHC class 1 antigen; MHC class I anti; MHC Class I antigen; MHC class I antigen A*30; MHC class I antigen A*33; MHC class I antigen A*74; MHC class I antigen heavy chain; MHC class I antigen HLA-A heavy chain; MHC class I antigen HLA-A33; MHC class I antigen null protein; MHC class I antigene; MHC class II antigen; MHC classI antigen; truncated MHC class I antigen | |
IgG2a κ | |
Affinity chromatography | |
HLA-A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title