Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA ABC Antibody (246-B8.E7), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311424G
Description
HLA ABC Monoclonal antibody specifically detects HLA ABC in Human, Mouse, Primate samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
HLA ABC | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Primate | |
Purified |
Flow Cytometry, Immunofluorescence | |
246-B8.E7 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80 | |
Human peripheral blood leukocytes | |
0.1 mL | |
Adaptive Immunity, Cell Biology, Immunology | |
3105 | |
Store at 4°C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction