Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA ABC Antibody (YTH862.2), Alexa Fluor™ 405, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB10065411AF405
Description
HLA ABC Monoclonal specifically detects HLA ABC in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
HLA ABC | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
HLA-A | |
PHA activated human peripheral blood lymphocytes. | |
0.1 mL | |
Adaptive Immunity, Cell Biology, Immunology | |
3105 | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
YTH862.2 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80 | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
NB100-65411 recognizes a monomorphic determinant on the human HLA Class I heavy chain. Epitope analysis has shown that this antibody binds in the alpha1 domain. MHC Class I is expressed by all nucleated cells. Recent studies have shown that this antibody may be able to induce apoptosis in activated but not resting T cells (1). | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction