Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA ABC Antibody (YTH862.2), Allophycocyanin/Cy7, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB10065411APCCY7
Description
HLA ABC Monoclonal specifically detects HLA ABC in Human samples. It is validated for Flow Cytometry.Specifications
HLA ABC | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
HLA-A | |
PHA activated human peripheral blood lymphocytes. | |
0.1 mL | |
Primary | |
NB100-65411 recognizes a monomorphic determinant on the human HLA Class I heavy chain. Epitope analysis has shown that this antibody binds in the alpha1 domain. MHC Class I is expressed by all nucleated cells. Recent studies have shown that this antibody may be able to induce apoptosis in activated but not resting T cells (1). | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
YTH862.2 | |
Flow Cytometry | |
A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80 | |
Rat | |
Protein G purified | |
Adaptive Immunity, Cell Biology, Immunology, Major Histocompatibility Complexes | |
3105.0 | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction