Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HLA-ABCE Monoclonal Antibody (TP25.99SF), PE
GREENER_CHOICE

Catalog No. PIMA544116
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544116 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544116 Supplier Invitrogen™ Supplier No. MA544116
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

This antibody recognizes an extracellular epitope on HLA-ABC and HLA-E molecules, but not HLA-G. It can be used for discrimination between HLA-G and other HLA-class I antigens.

HLA-A, -B, and -C are major histocompatibility complex (MHC)-class I antigens. Like other class I molecules (i.e. HLA-E, -F, -G), HLA-A, -B, and -C are heterodimers consisting of a 40 - 45 kDa transmembrane glycoproteic alpha-chain, noncovalently combined to the invariant B2-microglobulin. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-ABCE
Applications Flow Cytometry
Classification Monoclonal
Clone TP25.99SF
Concentration 0.1 mg/mL
Conjugate PE
Formulation PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
Gene HLA-B
Gene Accession No. P01889, P04439, P10321, P13747
Gene Alias PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
Gene Symbols HLA-A, HLA-B, HLA-C, HLA-E
Host Species Mouse
Immunogen IFN-gamma-treated human melanoma cells Colo 38.
Purification Method Size-exclusion chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3105, 3106, 3107, 3133
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.