Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-B Rat anti-Human, Alexa Fluor™ 647, Clone: YTH 76.3.rMAb (also known as YTH/76.3; YT76), BD Phrmingen™

Catalog No. BDB569662 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB569662 100 Tests
BDB569663 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB569662 Supplier BD Biosciences Supplier No. 569662
Add to Cart
Add to Cart

Rat Monoclonal Antibody

HLA-B Monoclonal antibody specifically detects HLA-B in Human samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-B
Applications Flow Cytometry
Classification Monoclonal
Clone YTH 76.3.rMAb (also known as YTH/76.3; YT76)
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias HLA-B; HLAB
Host Species Rat
Immunogen Normal Human Lymphocytes
Purification Method Affinity Chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3106
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.