Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA-B Rat anti-Human, BB700, Clone: YTH 76.3.rMAb (also known as YTH/76.3; YT76), BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 752626
This item is not returnable.
View return policy
Description
HLA-B Monoclonal antibody specifically detects HLA-B in Human samples. It is validated for Flow Cytometry
Specifications
HLA-B | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG1 κ |
Flow Cytometry | |
YTH 76.3.rMAb (also known as YTH/76.3; YT76) | |
Brilliant Blue 700 (BB700) | |
HLA-B; HLAB | |
Normal Human Lymphocytes | |
RUO | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction