Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA-B7 Monoclonal Antibody (BB7.1), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$417.00
Specifications
Antigen | HLA-B7 |
---|---|
Clone | BB7.1 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
25-591-742
![]() |
Invitrogen™
25591742 |
100 Tests |
Each of 1 for $417.00
|
|
|||||
Description
Mouse Monoclonal Antibody
HLA-B7 is one of the polymorphic proteins of human MHC Class I found on all nucleated cells. This surface protein is a heterodimer consisting of the specific transmembrane HLA-B chain paired with Beta-2 microglobulin. Classical MHC Class I molecules play a central role in adaptive immunity by presenting to T cells antigenic peptides derived either from the cell itself (classical presentation) or extracellular antigens (cross-presentation). The ligands of MHC class I include T cell receptor (TCR) and a costimulatory molecule CD8. Hundreds of HLA-B alleles have been described. Typing for these polymorphisms is routinely done for transplantation. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.Specifications
HLA-B7 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P01889 | |
HLA-B | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
BB7.1 | |
Flow Cytometry | |
PE-Cyanine7 | |
Mouse | |
Human | |
3106 | |
Papain solubilized HLA-B7. | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
HLA-B |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title