Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HLA-B7 Monoclonal Antibody (BB7.1), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 46591742
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-591-742 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 46-591-742 Supplier Invitrogen™ Supplier No. 46591742
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA-B7 is one of the polymorphic proteins of human MHC Class I found on all nucleated cells. This surface protein is a heterodimer consisting of the specific transmembrane HLA-B chain paired with Beta-2 microglobulin. Classical MHC Class I molecules play a central role in adaptive immunity by presenting to T cells antigenic peptides derived either from the cell itself (classical presentation) or extracellular antigens (cross-presentation). The ligands of MHC class I include T cell receptor (TCR) and a costimulatory molecule CD8. Hundreds of HLA-B alleles have been described. Typing for these polymorphisms is routinely done for transplantation. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-B7
Applications Flow Cytometry
Classification Monoclonal
Clone BB7.1
Concentration 5 μL/Test
Conjugate PerCP-eFluor 710
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene HLA-B
Gene Accession No. P01889
Gene Alias A29 protein; antigen presenting molecule; AS; Aw-33; Aw-74; B-4901; B-5001; Bw-47; Bw-50; CDABP0067; class I MHC, HLA-B antigen; DAQB-90C11.16; epididymis secretory protein Li 83; FLJ26655; HEL-S-83; histocompatibility antigen; HLA A; HLA -B; HLA B-40011; HLA B44; hla b7; HLA class I antigen; HLA class I antigen HLA-B; HLA class I histocompatibility antigen, A alpha chain; HLA class I histocompatibility antigen, A-1 alpha chain; HLA class I histocompatibility antigen, A-30 alpha chain; HLA class I histocompatibility antigen, A-33 alpha chain; HLA class I histocompatibility antigen, A-74 alpha chain; HLA class I histocompatibility antigen, B alpha chain; HLA class I histocompatibility antigen, B-47 alpha chain; HLA class I histocompatibility antigen, B-50 alpha chain; HLA class I histocompatibility antigen, B-82 alpha chain; HLA locus; HLA molecule; HLAA; HLA-A; HLA-A locus alpha 2 domain; HLA-A null; HLA-A11; HLA-A33; HLA-Aw33.1; HLAB; HLA-B; HLA-B 1502; HLA-B alpha chain; HLA-B alpha chain (B*2706); HLA-B alpha chain (B*5002); HLA-B alpha-chain; HLA-B*45ZJ; HLA-B15; HLA-B35; HLA-B35 antigen; HLA-B-3506; HLA-B39; HLA-B-3905; HLA-B49; HLA-B50; HLA-B55; HLA-B-5502; HLA-B-5602; HLA-B59; HLA-B61; HLA-Bw50 antigen; HLA-Cw; HLA-DQB1; HLA-DRB1; Human leukocyte antigen A; hypothetical protein; lecocyte antigen B; Leukocyte Antigen; leukocyte antigen class I-A; leukocyte antigen class I-B; Leukoocyte Antigen; lymphocyte antigen; major histocompatability complex B antigen; major histocompatibility complex, class I, A; major histocompatibility complex, class I, B; MCH class I antigen; MHC 1; MHC antigen; MHC class 1 antigen; MHC class I alpha chain; MHC class I anitgen; MHC class I anti; MHC Class I antigen; MHC class I antigen A*30; MHC class I antigen A*33; MHC class I antigen A*74; MHC class I antigen B*47; MHC class I antigen B*50; MHC class I antigen B*82; MHC class I antigen GN00104; MHC class I antigen heavy chain; MHC class I antigen HLA-A heavy chain; MHC class I antigen HLA-A33; MHC class I antigen HLA-B alpha chain; MHC class I antigen HLA-B heavy chain; MHC class I antigen null protein; MHC class I antigen SHCHA; MHC class I antigene; MHC class I human leukocyte antigen; MHC class I molecule; MHC class I protein; MHC class II antigen; MHC class l antigen; MHC classI antigen; MHC HLA-B cell surface glycoprotein; MHC HLA-B transmembrane glycoprotein; MHC HLA-B27d; MHC HLA-B27-HS; MHC HLA-B39N; MHC1; SPDA1; truncated class I antigen; truncated MHC class I antigen
Gene Symbols HLA-B
Host Species Mouse
Immunogen Papain solubilized HLA-B7.
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3106
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.