Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA-B7 Mouse anti-Human, BUV395, Clone: BB7.1, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 747889
Description
HLA-B7 Monoclonal antibody specifically detects HLA-B7 in Human samples. It is validated for Flow Cytometry.
Specifications
HLA-B7 | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
HLA-B7 | |
Refrigerate (2 to 8°C) |
Flow Cytometry | |
BB7.1 | |
Brilliant Ultraviolet 395 | |
50 μg | |
Primary | |
Human | |
IgG1 κ |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction