Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-C Mouse anti-Human, PE, Clone: DT-9 (also known as DT9), BD

Catalog No. BDB566372 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566372 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566372 Supplier BD Biosciences Supplier No. 566372
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

The DT-9 monoclonal antibody specifically recognizes Human Leukocyte Antigen (HLA-C), a polymorphic major histocompatibility complex (MHC) class I antigen. HLA-C is a heterodimer comprised of the HLA-C alpha chain, a ñ45 kDa type I transmembrane glycoprotein, and a ñ12 kDa Beta-2 (β2)-microglobulin light chain. HLA-C is expressed on nearly all cells, and plays a role in the antigen-specific, MHC-restricted presentation of small peptides to CD8+ T cells in the generation of immunity or tolerance. HLA-C may also bind to regulatory MHC class I antigen-selective receptors expressed by CD8+ T cells and natural killer (NK) cells. The DT-9 antibody also recognizes some rare HLA-A and HLA-B allotypes and the nonclassical HLA-E molecule.

R-phycoerythrin (PE) is an accessory photosynthetic pigment found in red algae. It exists in vitro as a 240-kDa protein with 23 phycoerythrobilin chromophores per molecule. This makes PE one of the brightest fluorochromes for flow cytometry applications, but its photobleaching properties make it unsuitable for fluorescence microscopy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-C
Classification Monoclonal
Clone DT-9 (also known as DT9)
Conjugate PE
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Gene Alias HLAC, HLC-C, HLA-JY3, MHC class I antigen C, PSORS1, HLA-E
Host Species Mouse
Immunogen Purified Human Tamarin MHC Class I Molecules
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Type Antibody
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.