Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HLA-C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587657
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587657 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587657 Supplier Invitrogen™ Supplier No. PA587657
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: LO2, HL-60, BT-474, Jurkat, Mouse liver; Cellular Location: Membrane, Single-pass type I membrane protein Immunogen sequence: CSHSMKYFFT SVSRPGRGEP RFISVGYVDD TQFVRFDSDA ASPRGEPRAP WVEQEGPEYW DRETQKYNRQ AQTDRVSLRN LRGYYNQSEA GSHTLQWMCG CDLGPDGRLL RGYDQYAYDG KDYIALNEDL RSWTAADTAA QITQRKWEAA REAEERRAYL EGTCVEWLRR YLENGKESLQ RAEHPKTHVT HHPVSDHEAT LRCWALGFYP AEITLTWQWD GEDQTQDTEL VETRPAGDGT FQKWAAVMVP SGEEQRYTCH VQHEGLPEPL TLRWEPSSQP TIPI.

HLA-C belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. Class I molecules play a central role in the immune system by presenting peptides derived from endoplasmic reticulum lumen. They are expressed in nearly all cells. The heavy chain is approximately 45 kDa and its gene contains 8 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the alpha1 and alpha2 domain, which both bind the peptide, exon 4 encodes the alpha3 domain, exon 5 encodes the transmembrane region, and exons 6 and 7 encode the cytoplasmic tail. Polymorphisms within exon 2 and exon 3 are responsible for the peptide binding specificity of each class one molecule. Typing for these polymorphisms is routinely done for bone marrow and kidney transplantation. Over one hundred HLA-C alleles have been described. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-C
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HLA-C
Gene Accession No. P10321
Gene Alias alpha chain of MHC class II; D6S204; HLA C; HLA class I histocompatibility antigen C alpha chain; HLA class I histocompatibility antigen Cw-1 alpha chain; HLA class I histocompatibility antigen, C alpha chain; HLA class I histocompatibility antigen, Cw-1 alpha chain; HLA class I histocompatibility antigen, Cw-15 alpha chain; HLA class I histocompatibility antigen, Cw-18 alpha chain; HLAC; HLA-C; HLA-C alpha chain; HLA-C alpha chain (HLA-Cw*15v); HLA-C alpha chain for Cw*18GB; HLA-C alpha-chain; HLA-C antigen; HLA-Cw; HLA-Cw*04GB; HLA-Cw*1702; HLA-Cw*18GB; HLA-Cw-1503; HLA-JY3; HLC-C; Human leukocyte antigen C; human leukocyte antigen-C alpha chain; leukocyte antigen C; major histocompatibility antigen HLA-C; major histocompatibility complex, class I, C; MHC; MHC clas I antigen; MHC class 1 antigen; MHC Class I antigen; MHC class I antigen Cw*15; MHC class I antigen Cw*18; MHC class I antigen heavy chain HLA-C; MHC class I histocompatibility antigen; MHC class I molecule; MHC class II antigen; PSORS1; surface antigen; truncated MHC class I antigen
Gene Symbols HLA-C
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 25-308 of human HLA-C (P30499).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3107
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.