Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HLA-C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119267
This item is not returnable.
View return policy
Description
HLA-C Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
HLA-C belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. Class I molecules play a central role in the immune system by presenting peptides derived from endoplasmic reticulum lumen. They are expressed in nearly all cells. The heavy chain is approximately 45 kDa and its gene contains 8 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the alpha1 and alpha2 domain, which both bind the peptide, exon 4 encodes the alpha3 domain, exon 5 encodes the transmembrane region, and exons 6 and 7 encode the cytoplasmic tail. Polymorphisms within exon 2 and exon 3 are responsible for the peptide binding specificity of each class one molecule. Typing for these polymorphisms is routinely done for bone marrow and kidney transplantation. Over one hundred HLA-C alleles have been described. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
Specifications
HLA-C | |
Polyclonal | |
Unconjugated | |
HLA-C | |
alpha chain of MHC class II; D6S204; HLA C; HLA class I histocompatibility antigen C alpha chain; HLA class I histocompatibility antigen Cw-1 alpha chain; HLA class I histocompatibility antigen, C alpha chain; HLA class I histocompatibility antigen, Cw-1 alpha chain; HLA class I histocompatibility antigen, Cw-15 alpha chain; HLA class I histocompatibility antigen, Cw-18 alpha chain; HLAC; HLA-C; HLA-C alpha chain; HLA-C alpha chain (HLA-Cw*15v); HLA-C alpha chain for Cw*18GB; HLA-C alpha-chain; HLA-C antigen; HLA-Cw; HLA-Cw*04GB; HLA-Cw*1702; HLA-Cw*18GB; HLA-Cw-1503; HLA-JY3; HLC-C; Human leukocyte antigen C; human leukocyte antigen-C alpha chain; leukocyte antigen C; major histocompatibility antigen HLA-C; major histocompatibility complex, class I, C; MHC; MHC clas I antigen; MHC class 1 antigen; MHC Class I antigen; MHC class I antigen Cw*15; MHC class I antigen Cw*18; MHC class I antigen heavy chain HLA-C; MHC class I histocompatibility antigen; MHC class I molecule; MHC class II antigen; PSORS1; surface antigen; truncated MHC class I antigen | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
3107 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
P10321 | |
HLA-C | |
Recombinant protein Major Histocompatibility Complex Class I C. The antigen corresponds to amino acid range 48-305 of the target protein. | |
200 μL | |
Primary | |
Human, Pig | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction