Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA Class I Antibody (H58A), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP260980
Description
HLA Class I Monoclonal specifically detects HLA Class I in Human, Bovine, Canine, Equine, Feline, Goat, Hamster, Llama, Ferret, Alpaca, Bison, Mink, Water Buffalo, Rabbit, Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
HLA Class I | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry 1:10 - 1:1000 | |
A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80 | |
Mouse | |
Unpurified | |
Primary | |
Human, Bovine, Canine, Equine, Feline, Goat, Hamster, Llama, Ferret, Alpaca, Bison, Mink, Water Buffalo, Rabbit, Sheep | |
IgG2a |
Flow Cytometry, Immunohistochemistry, Functional Assay | |
H58A | |
Unconjugated | |
Ascites with 0.09% Sodium Azide | |
HLA-A | |
Leukocytes from: horse, rabbit, rat, dog, goat,human | |
0.1 mg | |
3105 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction