Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA Class I Antibody (mAb90), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals DDX0250P100
Description
HLA Class I Monoclonal specifically detects HLA Class I in Human samples. It is validated for Flow Cytometry, ELISA, Functional.Specifications
HLA Class I | |
Monoclonal | |
0.5 mg/mL | |
Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Functional | |
HLA-A | |
HLA-A1 was used as immunogen. | |
0.1 mg | |
Adaptive Immunity, Cell Biology, Immunology | |
3105 | |
Human | |
IgG1 |
Flow Cytometry, ELISA, Functional Assay | |
mAb90 | |
Unconjugated | |
A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80 | |
Mouse | |
Ion exchange chromatography | |
RUO | |
Primary | |
human alpha1domain of HLA class I molecule | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction