Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA Class I Antibody (MEM-81), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500412AF594
Description
HLA Class I Monoclonal specifically detects HLA Class I in Human samples. It is validated for Flow Cytometry.Specifications
HLA Class I | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
HLA-A | |
Leukocytes of a patient suffering with LGL type leukemia. | |
Primary | |
The antibody MEM-81 reacts with human classical MHC Class I molecules in native cell-surface forms. MHC Class I molecules (MHC Class Ia) are expressed on the surface of human nucleated cell types. | |
Store at 4°C in the dark. |
Flow Cytometry | |
MEM-81 | |
Flow Cytometry | |
A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80 | |
Mouse | |
0.1 mL | |
3105 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction