Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA Class I Antibody (W6/32), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB7098J
This item is not returnable.
View return policy
Description
HLA Class I Monoclonal antibody specifically detects HLA Class I in Human samples. It is validated for Flow CytometrySpecifications
HLA Class I | |
Monoclonal | |
Janelia Fluor 646 | |
A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80 | |
Membranes from human tonsillar lymphocytes, | |
0.1 mL | |
Adaptive Immunity, Cell Biology, Immunology | |
3105 | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry | |
W6/32 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction