Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA Class I Heavy Chain (Restricted expression) Mouse anti-Human, Clone: HCA2, MUbio
SDP

Catalog No. 502101173
Encompass
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-210-1173 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-210-1173 Supplier Vector Laboratories Supplier No. MUB2036P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

HLA Class I Heavy Chain (Restricted expression) Monoclonal antibody specifically detects HLA Class I Heavy Chain (Restricted expression) in Human samples. It is validated for ELISA, Flow Cytometry, Immunoblot, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation, Western Blot

Specifications

Antigen HLA Class I Heavy Chain (Restricted expression)
Applications ELISA, Flow Cytometry, Immunoblot, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone HCA2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH
Gene HLA-A
Gene Accession No. P04439
Host Species Mouse
Immunogen Human HLA Class I Heavy Chain (Restricted expression)
Purification Method Protein A/G
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3105
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.