Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HLA-DMA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552978
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552978 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552978 Supplier Invitrogen™ Supplier No. PA552978
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PTPMWPDDLQ NHTFLHTVYC QDGSPSVGLS EAYDEDQLFF FDFSQNTRVP RLPEFADWAQ EQGDAPAILF DKEFCEWMIQ QIGPKLDGKI PVSRGFPIAE V Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 68%.

HLA-DMA belongs to the HLA class II alpha chain paralogues. This class II molecule is a heterodimer consisting of an alpha (DMA) and a beta chain (DMB), both anchored in the membrane. It is located in intracellular vesicles. DM plays a central role in the peptide loading of MHC class II molecules by helping to release the CLIP molecule from the peptide binding site. Class II molecules are expressed in antigen presenting cells (APC: B lymphocytes, dendritic cells, macrophages). The alpha chain is approximately 33-35 kDa and its gene contains 5 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the two extracellular domains, exon 4 encodes the transmembrane domain and the cytoplasmic tail. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-DMA
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HLA-DMA
Gene Accession No. P28067
Gene Alias class II histocompatibility antigen, M alpha chain; D6S222E; DAAP-27A1.5; DMA; HLA class II histocompatibility antigen, DM alpha chain; HLADM; HLA-DMA; major histocompatibility complex, class II, DM alpha; MHC class II antigen DMA; really interesting new gene 6 protein; RING6
Gene Symbols HLA-DMA
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HLA-DMA. Recombinant protein control fragment (Product #RP-90177).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3108
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.