Learn More
HLA-DMA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 118381AP150UL

Description
This antibody recognize the 33 kDa HLA-DMA and the 50-60 kDa dimer in Western blotting.
HLA-DMA belongs to the HLA class II alpha chain paralogues. This class II molecule is a heterodimer consisting of an alpha (DMA) and a beta chain (DMB), both anchored in the membrane. It is located in intracellular vesicles. DM plays a central role in the peptide loading of MHC class II molecules by helping to release the CLIP molecule from the peptide binding site. Class II molecules are expressed in antigen presenting cells (APC: B lymphocytes, dendritic cells, macrophages). The alpha chain is approximately 33-35 kDa and its gene contains 5 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the two extracellular domains, exon 4 encodes the transmembrane domain and the cytoplasmic tail.Specifications
HLA-DMA | |
Polyclonal | |
Unconjugated | |
HLA-DMA | |
D6S222E, DAMA 147C13.1 001, DAMC 226H3.5 001, DAQB 96B10.1 001, DASS 67H14.8 001, DMA, DMA protein, hCG_2001572, HLA DMA, HLA DMA protein, HLADM, HLA-DMA, RING6, XXbac BCX136A3.1 001, XXbac BPG181M17.3 001 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3108 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P28067 | |
HLA-DMA | |
HLA-DMA Fusion Protein Ag2452 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.