Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA DMB Antibody (SR1484), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | HLA DMB |
---|---|
Clone | SR1484 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50 |
Applications | Western Blot, Flow Cytometry |
Classification | Monoclonal |
Description
HLA DMB Monoclonal antibody specifically detects HLA DMB in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
HLA DMB | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
class II histocompatibility antigen, M beta chain, D6S221E, DMB, major histocompatibility complex, class II, DM beta, MHC class II antigen DMB, MHC class II antigen HLA-DM beta chain, MHC class II HLA-DMB, Really interesting new gene 7 protein, RING7HLA class II histocompatibility antigen, DM beta chain | |
A synthesized peptide derived from human HLA DMB (Uniprot #: P28068) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1484 | |
Western Blot, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Asthma, Diabetes Research, Immunology | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
3109 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title