Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HLA-DMB Recombinant Rabbit Monoclonal Antibody (3C2X4)
GREENER_CHOICE

Catalog No. PIMA542638
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542638 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542638 Supplier Invitrogen™ Supplier No. MA542638
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: HT-29, Raji. The target is usually found in the following locations: Late endosome membrane, Lysosome membrane, Single-pass type I membrane protein. Immunogen sequence: MITFLPLLLG LSLGCTGAGG FVAHVESTCL LDDAGTPKDF TYCISFNKDL LTCWDPEENK MAPCEFGVLN SLANVLSQHL NQKDTLMQRL RNGLQNCATH.

HLA-DMB (major histocompatibility complex, class II, DM beta), also known as D6S221E, RING7, HLA-DM histocompatibility type, beta chain, HLADMB or RING7, is a protein that in humans is encoded by the HLA-DMB gene. The HLA-DMB gene is mapped on 6p21.32. HLA-DMB belongs to the HLA class II beta chain paralogues. This class II molecule is a heterodimer consisting of an alpha (DMA) and a beta (DMB) chain, both anchored in the membrane. It is located in intracellular vesicles. DM plays a central role in the peptide loading of MHC class II molecules by helping to release the CLIP (class II-associated invariant chain peptide) molecule from the peptide binding site. Class II molecules are expressed in antigen presenting cells. The beta chain is approximately 26-28 kDa and its gene contains 6 exons. HLA-DMA and -DMB appear to encode subunits of a functional heterodimer that is critical in the pathway of class II antigen presentation. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-DMB
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 3C2X4
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HLA-DMB
Gene Accession No. P28068
Gene Alias class II histocompatibility antigen, M beta chain; D6S221E; DAAP-27A1.4; DMB; HLA class II histocompatibility antigen, DM beta chain; HLA-DMB; major histocompatibility complex, class II, DM beta; MHC class II antigen DMB; MHC class II antigen HLA-DM beta chain; MHC class II HLA-DMB; really interesting new gene 7 protein; RING7
Gene Symbols HLA-DMB
Host Species Rabbit
Immunogen Synthetic Peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3109
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.