Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HLA-DOA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560904
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560904 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560904 Supplier Invitrogen™ Supplier No. PA560904
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GDFARFDPQG GLAGIAAIKA HLDILVERSN RSRAINVPPR VTVLPKSRVE Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 70%.

Reacts with HLA-DO, a heterodimer formed by DNalpha and DObeta subunits in B cells. DNalpha and DObeta are the products of the non-classical class II genes, HLA-DNA and HLA-DOB, respectively. DO forms tight complexes with DM in the endoplasmic reticulum and is thereby sorted to lysosomal vesicles during antigen processing and presentation. It is tightly associated with DM and it is selectively expressed on antigen-presenting cells, such as B cells, dendritic cells and thymic epithelial cells. DO can enhance the efficiency of peptide loading and has been found to stabilize DM at low pH, preserving its chaperon activity. DO-DM complexes are more efficient than DM in protecting empty DR molecules. Reports describe DO as a co-chaperone of DM. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-DOA
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HLA-DOA
Gene Accession No. P06340
Gene Alias HLA class II histocompatibility antigen, DO alpha chain; HLA-D0-alpha; HLA-DNA; HLA-DOA; HLADZ; HLA-DZA; lymphocyte antigen; major histocompatibility complex, class II, DN alpha; major histocompatibility complex, class II, DO alpha; MHC class II antigen DOA; MHC DN-alpha; MHC DZ alpha
Gene Symbols HLA-DOA
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HLA-DOA. Recombinant protein control fragment (Product #RP-100030).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3111
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.