Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA DOB Antibody (DOB 10E6/5), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250420H
Description
HLA DOB Monoclonal specifically detects HLA DOB in Human samples. It is validated for Western Blot, ELISA.Specifications
HLA DOB | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
HLA-DOB | |
C-terminal peptide MHC class II | |
0.1 mL | |
Asthma, Immunology | |
3112 | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, ELISA | |
DOB 10E6/5 | |
Western Blot, ELISA | |
DOB, FLJ57033, HLA class II histocompatibility antigen, DO beta chain, major histocompatibility complex, class II, DO beta, MHC class II antigen DOB | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction