Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-DP/HLA-DQ/HLA-DR Mouse anti-Human, Clone: CR3/43, Abnova™

Catalog No. 89034765 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-765 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-765 Supplier Abnova Corporation Supplier No. MAB14682
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against human HLA-DP/HLA-DQ/HLA-DR.

  • High specificity
  • High reactivity

Specifications

Antigen HLA-DP/HLA-DQ/HLA-DR
Applications Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone CR3/43
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human HLA-DP/HLA-DQ/HLA-DR.
Dilution Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS.
Gene Alias DPB1/HLA-DP1B
Gene Symbols HLA-DPB1
Host Species Mouse
Immunogen Cells from human tonsil.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3115/3117/3122
Test Specificity Reacts with a common epitope of human major histocompatibility class II (MHC II) antigens, HLA-DP, HLA-DQ and HLA-DR
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.