Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA DPA Antibody (TAL3C3), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250601AF350
Description
HLA DPA Monoclonal specifically detects HLA DPA in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
HLA DPA | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
HLA-DPA1 | |
Bristol 8 separated alpha chain preparation | |
0.1 mL | |
3113 | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
TAL3C3 | |
Western Blot, Immunohistochemistry | |
DP alpha 1 chain, HLA class II histocompatibility antigen, DP alpha chain, HLADP, HLA-SB alpha chain, major histocompatibility complex, class II, DP alpha 1, MHC class II antigen, MHC class II DP3-alpha, MHC class II DPA1, MHC class II HLA-DPA1 antigen, Primed lymphocyte test-1 | |
Mouse | |
Protein A purified | |
Primary | |
TAL 3C3 is reactive on fixed tissue, reacts to class II antigens and cross-reacts with smooth muscle. Not cytotoxic with rabbit complement. TAL 3C3 detects alpha chain only by western blot and not the dimer. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction