Learn More
Description
HLA-DPA1 belongs to the HLA class II alpha chain paralogues. This class II molecule is a heterodimer consisting of an alpha (DPA) and a beta (DPB) chain, both anchored in the membrane. It plays a central role in the immune system by presenting peptides derived from extracellular proteins.
- Major histocompatibility complex, class II, DP alpha 1
- Transfected cell line: 293T
- Plasmid: pCMV-HLA-DPA1 full-length
- Host: Human
- Theoretical molecular weight: 29.3kDa
- Storage buffer: 1x sample buffer (50mM Tris-HCl, 2% SDS, 10% glycerol, 300mM 2-mercaptoethanol, 0.01% bromophenol blue)
Western Blotting
Specifications
Specifications
| Cell Line | 293T |
| For Use With (Application) | Western Blot |
| Description | HLA-DPA1 293T Cell Transient Overexpression Lysate(Denatured) |
| Gene Accession No. | BC009956 |
| Molecular Weight (g/mol) | 29.3 |
| pH Range | 6.8 |
| Plasmid | pCMV-HLA-DPA1 full-length |
| Quality Control Testing | Transient overexpression cell Lysate was tested with Anti-HLA-DPA1 antibody (H00003113-B01) by western blotting |
| Quantity | 100 μL |
| Storage Buffer | 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.