Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HLA-DPA1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553682
Description
Immunogen sequence: RPTGEFMFEF DEDEMFYVDL DKKETVWHLE EFGQAFSFEA QGGLANIAIL NNNLNTLIQR SNHTQATNDP PEVTVFPKEP VELGQPNTLI CHIDKFFPPV LNVTWLCNGE LVTEGVAESL FLPRTDYSFH KFHYLTF Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 65%.
HLA-DPA1 belongs to the HLA class II alpha chain paralogues. This class II molecule is a heterodimer consisting of an alpha (DPA) and a beta (DPB) chain, both anchored in the membrane. It plays a central role in the immune system by presenting peptides derived from extracellular proteins. Class II molecules are expressed in antigen presenting cells (APC: B lymphocytes, dendritic cells, macrophages). The alpha chain is approximately 33-35 kDa and its gene contains 5 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the two extracellular domains, exon 4 encodes the transmembrane domain and the cytoplasmic tail. Within the DP molecule both the alpha chain and the beta chain contain the polymorphisms specifying the peptide binding specificities, resulting in up to 4 different molecules. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
Specifications
| HLA-DPA1 | |
| Polyclonal | |
| Unconjugated | |
| HLA-DPA1 | |
| DP(W3); DP(W4); HLA class II histocompatibility antigen, DP alpha 1 chain; HLADP; HLA-DP1A; HLA-DPA1; HLASB; HLA-SB alpha chain; major histocompatibility complex, class II, DP alpha 1; MHC class II antigen; MHC class II DP3-alpha; MHC class II DPA1; MHC class II HLA-DPA1 antigen; PLT1; Primed lymphocyte test-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3113 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P20036 | |
| HLA-DPA1 | |
| Recombinant protein corresponding to Human HLA-DPA1. Recombinant protein control fragment (Product #RP-90437). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction