Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA DPB1 Antibody (BRA-FB6), Allophycocyanin , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311554APC
Description
HLA DPB1 Monoclonal antibody specifically detects HLA DPB1 in Human samples. It is validated for Flow CytometrySpecifications
HLA DPB1 | |
Monoclonal | |
APC | |
DP beta 1 chain, DPB1, HLA class II histocompatibility antigen, DP(W4) beta chain, HLA DP14-beta chain, HLA-DP histocompatibility type, beta-1 subunit, HLA-DP1B, HLA-DPB, major histocompatibility complex class II HLA DPB1 protein, major histocompatibility complex, class II, DP beta 1, MHC class II antigen beta chain, MHC class II antigen DP beta 1 chain, MHC class II antigen DPB1, MHC class II antigen DPbeta1, MHC class II HLA-DP-beta, MHC HLA DPB1 | |
Non-T, non-B human acute lymphoblastic leukemia REH6 cell line | |
0.1 mL | |
Asthma, Diabetes Research, Immunology | |
3115 | |
Store at 4°C in the dark. | |
IgG2b κ |
Flow Cytometry | |
BRA-FB6 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction