Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-DPB1 Mouse anti-Human, Unconjugated, Clone: BraFB6, Abnova™

Catalog No. 89337719 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-719 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-719 Supplier Abnova Corporation Supplier No. MAB21226
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

HLA-DPB1 Monoclonal antibody specifically detects HLA-DPB1 in Human samples. It is validated for IHC-P, IF, Flow Cyt.

Specifications

Antigen HLA-DPB1
Applications Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone BraFB6
Conjugate Unconjugated
Description major histocompatibility complex, class II, DP beta 1
Gene HLA-DPB1
Gene Alias DPB1, HLA-DP1B
Host Species Mouse
Immunogen Non-T, non-B human acute lymphoblastic leukemia REH6 cell line
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3115
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.