Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HLA-DPB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582411
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582411 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582411 Supplier Invitrogen™ Supplier No. PA582411
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YNREEFVRFD SDVGEFRAVT ELGRPDEEYW NSQKDILEEK RAVPDRMCRH NYELGGPMTL QRRVQPRVNV SPSKKGPLQH HNLLVCHVTD FYPGSIQVRW FLNGQEETAG VVSTNLIRNG DWTF Highest antigen sequence indentity to the following orthologs: Rat - 69%, Mouse - 69%.

HLA-DPB belongs to the HLA class II beta chain paralogues. This class II molecule is a heterodimer consisting of an alpha (DPA) and a beta chain (DPB), both anchored in the membrane. It plays a central role in the immune system by presenting peptides derived from extracellular proteins. Class II molecules are expressed in antigen presenting cells (APC: B lymphocytes, dendritic cells, macrophages). The beta chain is approximately 26-28 kDa and its gene contains 6 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the two extracellular domains, exon 4 encodes the transmembrane domain and exon 5 encodes the cytoplasmic tail. Within the DP molecule both the alpha chain and the beta chain contain the polymorphisms specifying the peptide binding specificities, resulting in up to 4 different molecules. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-DPB1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HLA-DPB1
Gene Accession No. P04440
Gene Alias beta1 domain MHC class II HLA DPB; CD; CELIAC1; class II HLA beta chain; DC-1 alpha chain; DC-alpha; DPB1; DQ-A1; GSE; histocompatibility antigen HLA-DR alpha; HLA class II histocompatibility antigen, DP beta 1 chain; HLA class II histocompatibility antigen, DP(W4) beta chain; HLA class II histocompatibility antigen, DQ alpha 1 chain; HLA class II histocompatibility antigen, DQ(W3) alpha chain; HLA class II histocompatibility antigen, DR alpha chain; HLA DP14-beta chain; HLA-DCA; HLA-DP; HLA-DP histocompatibility type, beta-1 subunit; HLA-DP1B; HLA-DPB; HLA-DPB1; HLA-DQA; HLA-DQA1; HLA-DRA; HLA-DRA1; leucocyte antigen DQA1; leukocyte antigen alpha chain; major histocompatibility complex class II antigen beta chain; major histocompatibility complex, class II, DP beta 1; major histocompatibility complex, class II, DQ alpha 1; major histocompatibility complex, class II, DR alpha; MHC cell surface glycoprotein; MHC cla; MHC class II antigen; MHC class II antigen beta chain; MHC class II antigen DP beta 1 chain; MHC class II antigen DPB1; MHC class II antigen DPbeta1; MHC class II antigen DRA; MHC class II DQA1; MHC class II HLA-D alpha glycoprotein; MHC class II HLA-DP-beta-1; MHC class II HLA-DQ-alpha-1; MHC class II HLA-DRB1; MHC class II surface glycoprotein; MHC HLA DPB1; MHC HLA-DQ alpha; MLRW
Gene Symbols HLA-DPB1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HLA-DPB1. Recombinant protein control fragment (Product #RP-90436).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3115
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.