Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA DPB1 Rabbit anti-Human, Clone: HLA-DPB1/2862R, Novus Biologicals™
SDP

Catalog No. p-7233348 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB122918 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB122918 Supplier R&D Systems Supplier No. NBP30772220UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Monoclonal Antibody

HLA DPB1 Monoclonal specifically detects HLA DPB1 in Human samples. It is validated for ELISA/Flow Cytometry/Immunocytochemistry/Immunofluorescence.

Specifications

Antigen HLA DPB1
Applications ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone HLA-DPB1/2862R
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 1-2 ug/million cells, ELISA 2-4 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Alias DP beta 1 chain, DPB1, HLA class II histocompatibility antigen, DP(W4) beta chain, HLA DP14-beta chain, HLA-DP histocompatibility type, beta-1 subunit, HLA-DP1B, HLA-DPB, major histocompatibility complex class II HLA DPB1 protein, major histocompatibility complex, class II, DP beta 1, MHC class II antigen beta chain, MHC class II antigen DP beta 1 chain, MHC class II antigen DPB1, MHC class II antigen DPbeta1, MHC class II HLA-DP-beta, MHC HLA DPB1
Host Species Rabbit
Immunogen Non-T, non-B human acute lymphoblastic leukemia REH6 cell line
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Asthma, Diabetes Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3115
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.