Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA DQ Antibody (TAL 4.1), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002700AF405
Description
HLA DQ Monoclonal specifically detects HLA DQ in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
HLA DQ | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
HLA-DQA1 | |
Whole cells from the B line WT46 | |
0.1 mL | |
Adaptive Immunity, Asthma, Immunology | |
3117 | |
Human | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
TAL 4.1 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
CD, CELIAC1DQ alpha 1 chain, DC-1 alpha chain, DQ-A1, FLJ27088, FLJ27328, GSE, HLA class II histocompatibility antigen, DQ(W3) alpha chain, HLA-DCA, HLA-DQA, leucocyte antigen DQA1, leukocyte antigen alpha chain, major histocompatibility complex, class II, DQ alpha 1, MGC149527, MHC class II antigen, MHC class II DQA1, MHC class II HLA-D alpha glycoprotein, MHC class II HLA-DQ-alpha-1, MHC class II surface glycoprotein, MHC HLA-DQ alpha | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human Leukocyte Antigen DQ1 (includes DQ5 and 6) | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction