Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA DQ/DR/DP Antibody (Bu26) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$192.47 - $517.59
Specifications
| Antigen | HLA DQ/DR/DP |
|---|---|
| Clone | Bu26 |
| Concentration | 1.0 mg/mL |
| Dilution | Flow Cytometry 1:100-1:200 |
| Applications | Flow Cytometry |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017560
![]() |
Novus Biologicals
NB100641620.025MG |
0.025 mg |
Each for $192.47
|
|
|||||
NB10064162
![]() |
Novus Biologicals
NB10064162 |
0.1 mg |
Each for $517.59
|
|
|||||
Description
HLA DQ/DR/DP Monoclonal specifically detects HLA DQ/DR/DP in Human samples. It is validated for Flow Cytometry.Specifications
| HLA DQ/DR/DP | |
| 1.0 mg/mL | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Asthma, Immunology | |
| P01909 | |
| 3117 | |
| Made to Human HLA DP DQ DR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Bu26 | |
| Flow Cytometry 1:100-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD, CELIAC1DQ alpha 1 chain, DC-1 alpha chain, DQ-A1, FLJ27088, FLJ27328, GSE, HLA class II histocompatibility antigen, DQ(W3) alpha chain, HLA-DCA, HLA-DQA, leucocyte antigen DQA1, leukocyte antigen alpha chain, major histocompatibility complex, class II, DQ alpha 1, MGC149527, MHC class II antigen, MHC class II DQA1, MHC class II HLA-D alpha glycoprotein, MHC class II HLA-DQ-alpha-1, MHC class II surface glycoprotein, MHC HLA-DQ alpha | |
| HLA-DQA1 | |
| IgG1 | |
| Protein G purified | |
| NB100-64162 recognizes the beta chains of class II human major histocompatibility (MHC) antigens. The antibody reacts with DP, DQ, DR beta chains, which are expressed by antigen presenting cells, B- cells, monocytes and activated T lymphocytes. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title