Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HLA-DQ Monoclonal Antibody (CC158)
GREENER_CHOICE

Catalog No. PIMA528485
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528485 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528485 Supplier Invitrogen™ Supplier No. MA528485
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse anti Bovine MHC class II DQ antibody, clone CC158 recognizes Bovine MHC Class II DQ. MHC Class II molecules are constitutively expressed on antigen presenting cells such as dendritic cells, B lymphocytes, monocytes, macrophages, activated T lymphocytes and may be induced on a range of other cell types by interferon gamma.

HLA-DQ is one of the HLA (human leukocyte antigen) molecules belonging to class II of the major histocompatibility protein (MHC) surface proteins. HLA-DQ and the other MHC class II molecules are heterodimeric proteins consisting of an alpha (DQA) and a beta chain (DQB), both anchored in the membrane. They play a central role in the immune system by presenting peptides derived from extracellular proteins. MHC Class II molecules are expressed in antigen-presenting cells (APC: B lymphocytes, dendritic cells, macrophages) and present peptides endocytosed, digested, and loaded onto the proteins prior to their migration to the cell surface. Other human MHC class II molecules are HLA-DP, HLA-DM, HLA-DR, HLA-DOA, and HLA-DOB. HLA-DQ displays significant variability, particularly in the beta subunit within the region that affects the peptides bound and presented by the protein. Different classes of DQ based on similar beta subunits allow for the grouping of these variants into serotypes, which can often be recognized by antibodies raised against these subunits. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-DQ
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone CC158
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene BOLA-DQB
Gene Accession No. Q9TVC8
Gene Alias BoLA-DQA; BOLA-DQA1; BOLA-DQA2; BOLA-DQA2 protein; BOLA-DQA3; BOLA-DQA5; BOLA-DQB; CELIAC1; HLA class II antigen; HLA class II histocompatibility antigen, DQ beta 1 chain; HLA-DQB; HLA-DQB1; HLA-DRB1; IDDM1; lymphocyte antigen; major histocompatability complex class II; major histocompatibility complex, class II, DQ alpha 1 precursor; major histocompatibility complex, class II, DQ alpha 2; major histocompatibility complex, class II, DQ alpha 5; major histocompatibility complex, class II, DQ alpha, type 1; major histocompatibility complex, class II, DQ alpha, type 3; major histocompatibility complex, class II, DQ beta; major histocompatibility complex, class II, DQ beta 1; MHC class II antigen; MHC class II antigen DQB1; MHC class II antigen HLA-DQ-beta-1; MHC class II antigen precursor; MHC class II DQ beta chain; MHC class II DQA; MHC class II DQA precursor; MHC class II DQA1 precursor; MHC class II DQA2 (BoLA-DQA2); MHC class II DQA2 precursor; MHC class II DQB precursor; MHC class II HLA-DQ beta glycoprotein; MHC class2 antigen; MHC DQ beta; similar to MHC class II DQA2 (BoLA-DQA2); Unknown (protein for MGC:127488); Unknown (protein for MGC:137100)
Gene Symbols BOLA-DQA1, BOLA-DQA2, BOLA-DQA3, BOLA-DQA5, BOLA-DQB
Host Species Mouse
Purification Method Protein G
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282494, 282495, 282534, 282535, 282536
Target Species Bovine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.