Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA DQ1/DQ3 Antibody (HL-37) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500405
Description
HLA DQ1/DQ3 Monoclonal specifically detects HLA DQ1/DQ3 in Human, Porcine samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF-ready.Specifications
HLA DQ/DQ3 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000, Flow Cytometry 1 to 4 μg/mL, Immunoprecipitation 1:50, CyTOF-ready | |
HLA-DQA1 | |
Burkitt's lymphoma cell line Raj | |
0.1 mg | |
Adaptive Immunity, Asthma, Immunology | |
3117 | |
Human, Pig | |
IgG3 |
Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF | |
HL-37 | |
Unconjugated | |
CD, CELIAC1DQ alpha 1 chain, DC-1 alpha chain, DQ-A1, FLJ27088, FLJ27328, GSE, HLA class II histocompatibility antigen, DQ(W3) alpha chain, HLA-DCA, HLA-DQA, leucocyte antigen DQA1, leukocyte antigen alpha chain, major histocompatibility complex, class II, DQ alpha 1, MGC149527, MHC class II antigen, MHC class II DQA1, MHC class II HLA-D alpha glycoprotein, MHC class II HLA-DQ-alpha-1, MHC class II surface glycoprotein, MHC HLA-DQ alpha | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
The antibody HL-37 reacts with polymorphic determinant on human HLA-DQ1 and HLA-DQ3 molecules (recognized epitope was found on isolated beta chain of DQ1), but does not react with HLA-DQ2. DQ is the isotype of human MHC Class II molecules expressed on antigen-presenting cells (APC; dendritic cells, B lymphocytes, monocytes, macrophages). | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction