Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-DQA1 Rabbit anti-Human, Unconjugated, Clone: HLA-DQA1/2866R, Abnova™

Catalog No. 89337852 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-852 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-852 Supplier Abnova Corporation Supplier No. RAB00427
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

HLA-DQA1 Monoclonal antibody specifically detects HLA-DQA1 in Human samples. It is validated for IHC-P, IF, Flow Cyt.

Specifications

Antigen HLA-DQA1
Applications Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone HLA-DQA1/2866R
Conjugate Unconjugated
Description major histocompatibility complex, class II, DQ alpha 1
Gene HLA-DQA1
Gene Alias CD, CELIAC1, DQ-A1, FLJ27088, FLJ27328, GSE, HLA-DQA, MGC149527
Host Species Rabbit
Immunogen Original antibody is raised against recombinant protein corresponding to full length human HLA-DQ
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3117, 3118, 3119
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.