Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HLA-DQA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597242
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597242 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597242 Supplier Invitrogen™ Supplier No. PA597242
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EDIVADHVAS YGVNFYQSHG PSGQYTHEFD GDEEFYVDLE TKETVWQLPM FSKFISFDPQ SALRNMAVGK HTLEFMMRQS NSTAATNEVP EVTVFSKFPV TL; Positive Samples: Mouse liver, Mouse kidney, Rat kidney; Cellular Location: Cell membrane, Endoplasmic reticulum membrane, Endosome membrane, Golgi apparatus, Lysosome membrane, Single-pass type I membrane protein, trans-Golgi network membrane.

HLA-DQA2 belongs to the HLA class II alpha chain family. The encoded protein forms a heterodimer with a class II beta chain. It is located in intracellular vesicles and plays a central role in the peptide loading of MHC class II molecules by helping to release the CLIP molecule from the peptide binding site. Class II molecules are expressed in antigen presenting cells (B lymphocytes, dendritic cells, macrophages) and are used to present antigenic peptides on the cell surface to be recognized by CD4 T-cells. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-DQA2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.95 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Alias DQ alpha 2; DX ALPHA; DX alpha chain; DX-ALPHA; HLA class II histocompatibility antigen, DQ alpha 2 chain; HLA class II histocompatibility antigen, DQ(6) alpha chain; HLA DXA; HLA-DQA1; HLA-DQA2; HLA-DXA; major histocompatibility complex, class II, DQ alpha 2; MHC class II DQA2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 24-125 of human HLA-DQA2 (NP_0644401).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.