Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA DQW1 Antibody (Genox 3.53), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002702B
Description
HLA DQW1 Monoclonal specifically detects HLA DQW1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
HLA DQW1 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
HLA-DQB1 | |
Bristol 8 glycoprotein from the Bristol 8 B lymphoblastoid cell line. | |
0.1 mL | |
Asthma, Diabetes Research, Immunology | |
3119 | |
Human | |
Purified |
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
Genox 3.53 | |
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
CELIAC1, DC1, HLA class II histocompatibility antigen, DQ beta 1 chain, HLA-DQB, IDDM1, LB-E12, MB1, MHC class II antigen DQB1, MHC class II antigen HLA-DQ-beta-1, MHC class II DQ beta chain, MHC class II HLA-DQ beta glycoprotein, MT1 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human Leukocyte Antigen DQW1 (DC1 determinant of human HLA-DR antigens) | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction