Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA-DR Antibody (L243), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10077855PCP
Description
HLA-DR Monoclonal specifically detects HLA-DR in Human, Canine, Baboon, Primate samples. It is validated for Flow Cytometry, Flow (Cell Surface).Specifications
| HLA-DR | |
| Monoclonal | |
| PerCP | |
| P01903 | |
| HLA-DRA | |
| Human lymphoblastoid cell line (RPMI 8866). | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Predicted cross-reactivity with Chimpanzee, Baboon, Cynomolgus, Rhesus, Marmoset, Tamarin, Squirrel Monkey and Canine. | |
| Store at 4C in the dark. |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| L243 | |
| Flow Cytometry, Flow (Cell Surface) | |
| FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MLRW | |
| Mouse | |
| 28 kDa | |
| 0.1 mL | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Biology, Cellular Markers, Diabetes Research, Immunology, Major Histocompatibility Complexes | |
| 3122 | |
| Human, Canine, Baboon, Primate | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction