Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA-DR Antibody (TAL 8.1), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002708PEATT594
Description
HLA-DR Monoclonal specifically detects HLA-DR in Human samples. It is validated for Flow Cytometry.Specifications
HLA-DR | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
HLA-DRA | |
Cells from the B line WVB coated with mouse antiserum followed by WVB class II antigen. | |
0.1 mL | |
Primary | |
Specific for Human Leukocyte Antigens DR3, 6, 11, DRJ25. May be used for HLA typing. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
TAL 8.1 | |
Flow Cytometry | |
FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MLRW | |
Mouse | |
Protein A purified | |
Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Biology, Cellular Markers, Diabetes Research, Immunology, Major Histocompatibility Complexes | |
3122.0 | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction