Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA DR/DP Antibody (CL10962), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP321187100UL
Description
HLA DR/DP Monoclonal antibody specifically detects HLA DR/DP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
HLA DR/DP | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
CL10962 | |
Western Blot 1 ug/ml, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MLRW | |
This antibody was developed against a Synthetic Peptide of P01903. Actual immunogen sequence is proprietary. | |
100 μg | |
Adaptive Immunity, Cell Biology, Diabetes Research, Immunology | |
3122 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction