Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA DR/DP Antibody (MEM-136), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500447AF647
Description
HLA DR/DP Monoclonal specifically detects HLA DR/DP in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
HLA DR/DP | |
Monoclonal | |
Alexa Fluor 647 | |
FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MLRW | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
The antibody MEM-136 recognizes common epitope on beta-chain of human HLA-DR and HLA-DP. It reacts with alpha/beta dimer as well as with dissociated beta-subunit. DR and DP are the isotypes of human MHC Class II molecules expressed on antigen-presenting cells (APC; dendritic cells, B lymphocytes, monocytes, macrophages). | |
Store at 4C in the dark. |
Western Blot, Flow Cytometry | |
MEM-136 | |
Western Blot, Flow Cytometry | |
HLA-DRA | |
PHA-activated peripheral blood lymphocytes. | |
0.1 mL | |
Adaptive Immunity, Cell Biology, Diabetes Research, Immunology | |
3122 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction