Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA DR/DP Antibody (MEM-136), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500447PECY7
Description
HLA DR/DP Monoclonal specifically detects HLA DR/DP in Human samples. It is validated for Flow Cytometry.Specifications
HLA DR/DP | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
HLA-DRA | |
PHA-activated peripheral blood lymphocytes. | |
0.1 mL | |
Primary | |
The antibody MEM-136 recognizes common epitope on beta-chain of human HLA-DR and HLA-DP. It reacts with alpha/beta dimer as well as with dissociated beta-subunit. DR and DP are the isotypes of human MHC Class II molecules expressed on antigen-presenting cells (APC; dendritic cells, B lymphocytes, monocytes, macrophages). | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
MEM-136 | |
Flow Cytometry | |
FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MLRW | |
Mouse | |
Protein A purified | |
Adaptive Immunity, Cell Biology, Diabetes Research, Immunology | |
3122.0 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction