Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HLA-DR/DP/DQ Monoclonal Antibody (Bu26)
GREENER_CHOICE

Catalog No. PIMA184031
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA184031 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA184031 Supplier Invitrogen™ Supplier No. MA184031
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human HLA DP DQ DR antibody, clone Bu26 reacts with DP, DQ and DR beta chains, which are expressed by antigen presenting cells, B cells, monocytes and activated T lymphocytes.

MHC class II molecules are transmembrane glycoproteins composed of an alpha chain (36 kDa) and a beta chain (27 kDa). They are expressed primarily on antigen presenting cells such as B lymphocytes, monocytes, macrophages, thymic epithelial cells and activated T lymphocytes. Three loci, DR, DQ and DP, encode the major expressed products of the human class II region. The human MHC class II molecules bind intracellularly processed peptides and present them to T-helper cells. They therefore have a critical role in the initiation of the immune response. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-DR/DP/DQ
Applications Flow Cytometry
Classification Monoclonal
Clone Bu26
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene HLA-DRB5
Gene Accession No. P01903, P01911, P01920, P04440, P13762, P79483, Q30154
Gene Alias beta1 domain MHC class II HLA DPB; CELIAC1; class II HLA beta chain; Clone P2-beta-3; DLA class II histocompatibility antigen, DR-1 beta chain; DLA DRBB1 beta chain; DLA-DR beta; DLA-DRB1; DLA-DRBB1; DPB1; DR-16; DRB; DRB1; DRBB1; DRw10; DW2.2/DR2.2; FLJ75017; FLJ76359; HLA class II antigen; HLA class II histocompatibility antigen, DP beta 1 chain; HLA class II histocompatibility antigen, DP(W4) beta chain; HLA class II histocompatibility antigen, DQ beta 1 chain; HLA class II histocompatibility antigen, DR-1 beta chain; HLA class II histocompatibility antigen, DRB1-15 beta chain; HLA class II histocompatibility antigen, DRB1-16 beta chain; HLA class II histocompatibility antigen, DRB1-3 chain; HLA DP14-beta chain; HLA-D; HLA-DP; HLA-DP histocompatibility type, beta-1 subunit; HLA-DP1B; HLA-DPB; HLA-DPB1; HLA-DQ; HLA-DQB; HLA-DQB1; HLA-DR; HLA-DR1B; HLA-DRB; HLA-DRB1; HLA-DRB2; human leucocyte antigen DRB1; IDDM1; integral membrane glycoprotein; LA-DRB; lymphocyte antigen; lymphocyte antigen DRB1; major histocompatibility complex class II antigen beta chain; major histocompatibility complex, class II, DP beta 1; major histocompatibility complex, class II, DQ beta 1; major histocompatibility complex, class II, DR beta 1; MHC Class 2; MHC class II; MHC class II antigen; MHC class II antigen beta chain; MHC class II antigen DP beta 1 chain; MHC class II antigen DPB1; MHC class II antigen DPbeta1; MHC class II antigen DQB1; MHC class II antigen DRB1*15; MHC class II antigen DRB1*16; MHC class II antigen DRB1*3; MHC class II antigen HLA-DQ-beta-1; MHC class II DLA DRB1 beta chain; MHC class II DLA-DRB; MHC class II DLA-DR-beta-1; MHC class II DQ beta chain; MHC class II DR beta 1; MHC class II HLA-DP-beta-1; MHC class II HLA-DQ beta glycoprotein; MHC class II HLA-DR beta 1 chain; MHC class II HLA-DRB1; MHC class II HLA-DR-beta cell surface glycoprotein; MHC class II HLA-DRw10-beta; MHC class2 antigen; MHC DLA-DR-beta-1 precursor; MHC DQ beta; MHC HLA DPB1; MHC HLA-D; MHC II HLA; SS1; XXbac-BPG161M6.1
Gene Symbols HLA-DPB1, HLA-DQB1, HLA-DRA, HLA-DRB1, HLA-DRB3, HLA-DRB4, HLA-DRB5
Host Species Mouse
Immunogen Proprietary Immunogen.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3115, 3119, 3122, 3123, 3125, 3126, 3127
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.