Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HLA-DR/DP/DQ Monoclonal Antibody (CR3/43)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA125914

Catalog No. PIMA125914


Description

Description

Recommended positive controls: Tonsil.

MHC class II molecules are transmembrane glycoproteins composed of an alpha chain (36 kDa) and a beta chain (27 kDa). They are expressed primarily on antigen presenting cells such as B lymphocytes, monocytes, macrophages, thymic epithelial cells and activated T lymphocytes. Three loci, DR, DQ and DP, encode the major expressed products of the human class II region. The human MHC class II molecules bind intracellularly processed peptides and present them to T-helper cells. They therefore have a critical role in the initiation of the immune response. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HLA-DR/DP/DQ
Monoclonal
Conc. Not Determined
tissue culture supernatant with 0.09% sodium azide
P01903, P01911, P01920, P04440, P13762, P79483, Q30154
HLA-DPB1, HLA-DQB1, HLA-DRA, HLA-DRB1, HLA-DRB3, HLA-DRB4, HLA-DRB5
This information is considered to be commercially sensitive.
RUO
3115, 3119, 3122, 3123, 3125, 3126, 3127
4°C
Liquid
Immunohistochemistry (Paraffin)
CR3/43
Unconjugated
HLA-DRB5
beta1 domain MHC class II HLA DPB; CELIAC1; class II HLA beta chain; Clone P2-beta-3; DLA class II histocompatibility antigen, DR-1 beta chain; DLA DRBB1 beta chain; DLA-DR beta; DLA-DRB1; DLA-DRBB1; DPB1; DR-16; DRB; DRB1; DRBB1; DRw10; DW2.2/DR2.2; FLJ75017; FLJ76359; HLA class II antigen; HLA class II histocompatibility antigen, DP beta 1 chain; HLA class II histocompatibility antigen, DP(W4) beta chain; HLA class II histocompatibility antigen, DQ beta 1 chain; HLA class II histocompatibility antigen, DR-1 beta chain; HLA class II histocompatibility antigen, DRB1-15 beta chain; HLA class II histocompatibility antigen, DRB1-16 beta chain; HLA class II histocompatibility antigen, DRB1-3 chain; HLA DP14-beta chain; HLA-D; HLA-DP; HLA-DP histocompatibility type, beta-1 subunit; HLA-DP1B; HLA-DPB; HLA-DPB1; HLA-DQ; HLA-DQB; HLA-DQB1; HLA-DR; HLA-DR1B; HLA-DRB; HLA-DRB1; HLA-DRB2; human leucocyte antigen DRB1; IDDM1; integral membrane glycoprotein; LA-DRB; lymphocyte antigen; lymphocyte antigen DRB1; major histocompatibility complex class II antigen beta chain; major histocompatibility complex, class II, DP beta 1; major histocompatibility complex, class II, DQ beta 1; major histocompatibility complex, class II, DR beta 1; MHC Class 2; MHC class II; MHC class II antigen; MHC class II antigen beta chain; MHC class II antigen DP beta 1 chain; MHC class II antigen DPB1; MHC class II antigen DPbeta1; MHC class II antigen DQB1; MHC class II antigen DRB1*15; MHC class II antigen DRB1*16; MHC class II antigen DRB1*3; MHC class II antigen HLA-DQ-beta-1; MHC class II DLA DRB1 beta chain; MHC class II DLA-DRB; MHC class II DLA-DR-beta-1; MHC class II DQ beta chain; MHC class II DR beta 1; MHC class II HLA-DP-beta-1; MHC class II HLA-DQ beta glycoprotein; MHC class II HLA-DR beta 1 chain; MHC class II HLA-DRB1; MHC class II HLA-DR-beta cell surface glycoprotein; MHC class II HLA-DRw10-beta; MHC class2 antigen; MHC DLA-DR-beta-1 precursor; MHC DQ beta; MHC HLA DPB1; MHC HLA-D; MHC II HLA; SS1; XXbac-BPG161M6.1
Mouse
1 mL
Primary
Human
Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.